  • Search Results
  • Metabolic Glycoengineering of Cell-Derived Matrices and Cell Surfaces: A Combination of Key Principles and Step-by-Step Procedures.
Metabolic Glycoengineering of Cell-Derived Matrices and Cell Surfaces: A Combination of Key Principles and Step-by-Step Procedures.
ACS biomaterials science & engineering (2019-01-14)
Marcus Gutmann, Julian Bechold, Jürgen Seibel, Lorenz Meinel, Tessa Lühmann
ABSTRACT
Metabolic glycoengineering allows insertion of non-natural monosaccharides into glycan structures during biosynthesis thereby enabling extracellular matrices (ECMs), cell surfaces, or tissues for decoration with functional cues with ultimate spatial control while deploying aqueous and toxicologically benign coupling chemistries. In this work, we discuss relevant methods in the design of metabolic glycoengineered systems, ranging from synthetic procedures to decoration of cell surfaces and ECM components by bioorthogonal chemistries for widespread biomedical applications. As representative example, we chose a tetra-acetylated azide-bearing monosaccharide as model compound to be metabolically incorporated into glycans of the glycocalyx and ECM components generated by NIH 3T3 cells. Detailed guidance in fabrication and functionalization of azide-bearing glycan structures via bioorthogonal click chemistries in glycoengineered extracellular matrices is provided. In addition, a biocompatible design space of the copper(I)-catalyzed azide-alkyne cycloaddition due to the toxicity of the copper catalyst is detailed enabling effective and safe modification of living cell systems. Thereby, this set of methods provides the blueprint enabling the design and characterization of metabolically glycoengineered systems for novel applications in drug delivery and tissue engineering.
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Ponceau S, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
D-(+)-Glucosamine hydrochloride, ≥99%, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%