Careful selection of the host embryo is critical to the efficient production of knockout (KO) mice when injecting mouse embryonic stem (mES) cells into blastocysts. B6(Cg)-Tyrc-2j/J (B6 albino) and C57BL/6NTac (B6NTac) strains of mice are widely used to produce host blastocysts for such procedures. Here, we tested these two strains to identify an appropriate match for modified agouti C57BL/6N (JM8A3.N1) mES cells. When comparing blastocyst yield, super-ovulated B6NTac mice produced more injectable blastocysts per female than B6 albino mice (8.2 vs. 5.4). There was no significant difference in birth rate when injected embryos were transferred to the same pseudopregnant recipient strain. However, the live birth rate was significantly higher for B6NTac blastocysts than B6 albino blastocysts (62.7% vs. 50.2%). In addition, the proportion of pups exhibiting high-level and complete chimerism, as identified by coat color, was also significantly higher in the B6NTac strain. There was no obvious difference in the efficiency of germline transmission (GLT) when compared between B6NTac and B6 albino host embryos (61.5% vs. 63.3% for mES clones; 64.5% vs. 67.9% for genes, respectively), thus suggesting that an equivalent GLT rate could be obtained with only a few blastocyst injections for B6NTac embryos. In conclusion, our data indicate that B6NTac blastocysts are a better choice for the microinjection of JM8A3.N1 mES cells than B6 albino blastocysts.