  • Search Results
  • Targeting of neurotrophic factors, their receptors, and signaling pathways in the developmental neurotoxicity of organophosphates in vivo and in vitro.
Targeting of neurotrophic factors, their receptors, and signaling pathways in the developmental neurotoxicity of organophosphates in vivo and in vitro.
Brain research bulletin (2008-05-27)
Theodore A Slotkin, Frederic J Seidler, Fabio Fumagalli
ABSTRACT
Neurotrophic factors control neural cell differentiation and assembly of neural circuits. We previously showed that organophosphate pesticides differentially regulate members of the fibroblast growth factor (fgf) gene family. We administered chlorpyrifos and diazinon to neonatal rats on postnatal days 1-4 at doses devoid of systemic toxicity or growth impairment, and spanning the threshold for barely-detectable cholinesterase inhibition. We evaluated the impact on gene families for different classes of neurotrophic factors. Using microarrays, we examined the regional expression of mRNAs encoding the neurotrophins (ntfs), brain-derived neurotrophic factor (bdnf), nerve growth factor (ngf), the wnt and fzd gene families and the corresponding receptors. Chlorpyrifos and diazinon both had widespread effects on the fgf, ntf, wnt and fzd families but much less on the bdnf and ngf groups. However, the two organophosphates showed disparate effects on a number of key neurotrophic factors. To determine if the actions were mediated directly on differentiating neurons, we tested chlorpyrifos in PC12 cells, an in vitro model of neural cell development. Effects in PC12 cells mirrored many of those for members of the fgf, ntf and wnt families, as well as the receptors for the ntfs, especially during early differentiation, the stage known to be most susceptible to disruption by organophosphates. Our results suggest that actions on neurotrophic factors provide a mechanism for the developmental neurotoxicity of low doses of organophosphates, and, since effects on expression of the affected genes differed with test agent, may help explain regional disparities in effects and critical periods of vulnerability.
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Horse Serum, Donor herd, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Horse Serum, Donor Herd, USA origin, Heat inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Horse serum, liquid
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Horse Serum, USA origin, Donor herd, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma