  • Search Results
  • A distinct peripheral blood monocyte phenotype is associated with parasite inhibitory activity in acute uncomplicated Plasmodium falciparum malaria.
A distinct peripheral blood monocyte phenotype is associated with parasite inhibitory activity in acute uncomplicated Plasmodium falciparum malaria.
PLoS pathogens (2009-10-24)
Pattamawan Chimma, Christian Roussilhon, Panudda Sratongno, Ronnatrai Ruangveerayuth, Kovit Pattanapanyasat, Jean-Louis Pérignon, David J Roberts, Pierre Druilhe
ABSTRACT
Monocyte (MO) subpopulations display distinct phenotypes and functions which can drastically change during inflammatory states. We hypothesized that discrete MO subpopulations are induced during malaria infection and associated with anti-parasitic activity. We characterized the phenotype of blood MO from healthy malaria-exposed individuals and that of patients with acute uncomplicated malaria by flow cytometry. In addition, MO defense function was evaluated by an in vitro antibody dependent cellular inhibition (ADCI) assay. At the time of admission, the percentages and absolute numbers of CD16+ MO, and CCR2+CX3CR1+ MO, were high in a majority of patients. Remarkably, expression of CCR2 and CX3CR1 on the CD14(high (hi)) MO subset defined two subgroups of patients that also differed significantly in their functional ability to limit the parasite growth, through the ADCI mechanism. In the group of patients with the highest percentages and absolute numbers of CD14(hi)CCR2+CX3CR1+ MO and the highest mean levels of ADCI activity, blood parasitemias were lower (0.14+/-0.34%) than in the second group (1.30+/-3.34%; p = 0.0053). Data showed that, during a malaria attack, some patients' MO can exert a strong ADCI activity. These results bring new insight into the complex relationships between the phenotype and the functional activity of blood MO from patients and healthy malaria-exposed individuals and suggest discrete MO subpopulations are induced during malaria infection and are associated with anti-parasitic activity.
Product Description
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture