  • Search Results
  • myo-Inositol trispyrophosphate: a novel allosteric effector of hemoglobin with high permeation selectivity across the red blood cell plasma membrane.
myo-Inositol trispyrophosphate: a novel allosteric effector of hemoglobin with high permeation selectivity across the red blood cell plasma membrane.
Chembiochem : a European journal of chemical biology (2010-11-19)
Carolina D Duarte, Ruth Greferath, Claude Nicolau, Jean-Marie Lehn
ABSTRACT
myo-Inositol trispyrophosphate (ITPP), a novel membrane-permeant allosteric effector of hemoglobin (Hb), enhances the regulated oxygen release capacity of red blood cells, thus counteracting the effects of hypoxia in diseases such as cancer and cardiovascular ailments. ITPP-induced shifting of the oxygen-hemoglobin equilibrium curve in red blood cells (RBCs) was inhibited by DIDS and NAP-taurine, indicating that band 3 protein, an anion transporter mainly localized on the RBC membrane, allows ITPP entry into RBCs. The maximum intracellular concentration of ITPP, determined by ion chromatography, was 5.5×10(-3) M, whereas a drop in concentration to the limit of detection was observed in NAP-taurine-treated RBCs. The dissociation constant of ITPP binding to RBC ghosts was found to be 1.72×10(-5) M. All data obtained indicate that ITPP uptake is mediated by band 3 protein and is thus highly tissue-selective towards RBCs, a feature of major importance for its potential therapeutic use.
Sigma-Aldrich
2,3-Diphospho-D-glyceric acid pentasodium salt, glycolysis metabolite
Sigma-Aldrich
Phytase from wheat, ≥0.01 unit/mg solid
Sigma-Aldrich
4,4′-Diisothiocyanatostilbene-2,2′-disulfonic acid disodium salt hydrate, ≥80% (elemental analysis), powder
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%