  • Search Results
  • Pattern of expression of vascular endothelial growth factor and its receptors in the ovine choroid plexus during long and short photoperiods.
Pattern of expression of vascular endothelial growth factor and its receptors in the ovine choroid plexus during long and short photoperiods.
Cell and tissue research (2012-05-25)
Aleksandra Szczepkowska, Barbara Wąsowska, Przemysław D Gilun, Christine Lagaraine, Vincent Robert, Laurence Dufourny, Jean-Claude Thiéry, Janina Skipor
ABSTRACT
Vascular endothelial growth factor (VEGF-A) plays an important role in maintaining cerebrospinal fluid (CSF) homeostasis and the function of the choroid plexuses (CPs). The objective of the study was to determine the expression of vascular endothelial growth factor (VEGF-A), tyrosine kinase receptors Flt-1 and KDR and KDR co-receptor neuropilin 1 (NRP-1) in ovine CPs during different photoperiods. CPs were collected from the lateral brain ventricles from ovariectomized, estradiol-treated ewes during long day (LD; 16L:8D, n = 5) and short day (SD; 8L:16D, n = 5) photoperiods. We analyzed mRNA expression levels of two VEGF-A isoforms, VEGF-A (120) and VEGF-A (164) and our results indicate that VEGF-A (164) was the predominant isoform. Expression levels of VEGF-A and Flt-1 were similar during the SD and LD photoperiods. There were significant increases in KDR mRNA and protein expression (p < 0.05) and NRP-1 mRNA expression (p < 0.05) during SD. These data show that expression of KDR and its co-receptor NRP-1 are up-regulated by short photoperiod and that this effect is not dependent on ovarian steroids. Our results suggest that the VEGF-A-system may be involved in photoperiodic plasticity of CP capillaries and may therefore be responsible for photoperiodic changes in the CSF turnover rate in ewes.
Sigma-Aldrich
REDTaq® ReadyMix PCR Reaction Mix, Complete PCR reagent with standard Taq DNA Polymerase and inert dye