  • Search Results
  • Expression of BMP-7 in human gastric cancer and its clinical significance.
Expression of BMP-7 in human gastric cancer and its clinical significance.
British journal of cancer (2011-01-13)
M Aoki, S Ishigami, Y Uenosono, T Arigami, Y Uchikado, Y Kita, H Kurahara, M Matsumoto, S Ueno, S Natsugoe
ABSTRACT
Bone morphogenetic protein-7 (BMP-7) is a signalling molecule belonging to the transforming growth factor--superfamily. Recent studies have demonstrated the clinical impact of BMP-7 expression in various human cancers. However, there have been few reports detailing this in gastric cancer. We immunohistochemically investigated the expression of BMP-7 in 233 gastric cancer patients to disclose the clinicopathological features of BMP-7-positive gastric cancer. Immunohistochemically, in human gastric cancer, BMP-7 expression was identified in cellular membranes but also in the cytoplasm of cancer cells. Bone morphogenetic protein-7-positive expression was found in 129 of 233 patients (55%). Bone morphogenetic protein-7 expression was correlated with tumour size, nodal involvement, lymphatic invasion, venous invasion and histology (P<0.05). Bone morphogenetic protein-7 expression was significantly correlated with patient postoperative outcome, especially in the undifferentiated group. Multivariate analysis revealed BMP-7 expression as one of the independent prognostic factors next to the depth of invasion and nodal involvement (P<0.01). From the data collected, it would be appropriate to conclude on the possible regulation of gastric cancer progression by autocrine or paracrine BMP-7 loops. We can use BMP-7 expression as one of the strong predictors of risk of tumour recurrence in gastric cancer.
Product Description
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture