Merck
  • Prostaglandin DP receptors positively coupled to adenylyl cyclase in embryonic bovine tracheal (EBTr) cells: pharmacological characterization using agonists and antagonists.
Prostaglandin DP receptors positively coupled to adenylyl cyclase in embryonic bovine tracheal (EBTr) cells: pharmacological characterization using agonists and antagonists.
British journal of pharmacology (1999-06-16)
J Y Crider, B W Griffin, N A Sharif
ABSTRACT
Various prostaglandin agonists representing various classes of receptor subtypes were evaluated for their ability to stimulate adenylyl cyclase via the endogenous DP receptor in embryonic bovine tracheal (EBTr) cells. Two antagonists were used to block the agonist-induced cyclic AMP production. ZK118182 (EC50 = 16+/-4 nM), RS-93520 (EC50 = 23+/- 4 nM), SQ27986 (EC50 = 33+/-9 nM), ZK110841 (EC50 = 33+/-5 nM), BW245C (EC50 = 59+/-19 nM) and PGD2 (EC50=101+/-10 nM) (n = 4-70) were the most potent agonists. Whilst most compounds were full agonists (Emax = 100% relative to PGD2), BW245C was significantly more efficacious than PGD2 (Emax = 121+/-3%; P<0.001) and RS-93520 appeared to be a partial agonist (Emax = 64+/-9%; P<0.001). Agonists from the EP (e.g. enprostil; misoprostol; butaprost), FP (e.g. cloprostenol; fluprostenol; PHXA85), IP (iloprost; PGI2) and TP (U46619) prostanoid receptor classes were weak agonists or inactive in the EBTr cell assay system. The DP-receptor antagonist, BWA868C, showed a competitive antagonist profile with pA2 values of 8.00+/-0.02 and 8.14+/-0.13 in Schild analyses with two structurally different agonists, BW245C and ZK118182, respectively (n = 3). AH6809, another purported DP-receptor antagonist, weakly inhibited PGD2- and ZK 18182-induced cyclic AMP production (K(i)s = 808+/-193 nM and 782+/-178 nM, respectively). The current studies have characterized the DP receptor positively coupled to adenylyl cyclase in EBTr cells using a wide range of agonist and antagonist prostaglandins. These data support the utility of the EBTr cell line as a useful tool for the evaluation of DP receptor agonists and antagonists and for profiling other classes of prostaglandins.
Sigma-Aldrich
BW A868C, ≥98% (HPLC)
Sigma-Aldrich
AH6809, ≥98%, crystalline solid or supercooled liquid