  • Search Results
  • A polychromatic staining method for epoxy embedded tissue: a new combination of methylene blue and basic fuchsine for light microscopy.
A polychromatic staining method for epoxy embedded tissue: a new combination of methylene blue and basic fuchsine for light microscopy.
Biotechnic & histochemistry : official publication of the Biological Stain Commission (2006-05-25)
F D'Amico
ABSTRACT
A simple and rapid method is described for staining semithin sections of material embedded in epoxy resin for observing tissues prior to transmission electron microscopy. The method is suitable for tissue fixed with a glutaraldehyde-formaldehyde mixture and postfixed in osmium tetroxide. No etching or oxidizing procedures are necessary. Sections 0.5-0.8 microm thick are dried onto a slide and stained with either 0.75% methylene blue and 0.25% azure B or 0.5% methylene blue and 0.5% azure II in 0.5% aqueous borax and heated over a flame for 8-10 sec. The slides are rinsed with water, then stained the same way with 0.1% basic fuchsine in 5% aqueous ethanol. Cytoplasm stains blue; nuclei darker blue; collagen, mucus and elastin pink to red; fat and intracellular lipid droplets gray-green.
Sigma-Aldrich
Azure B, prepared by direct synthesis
Sigma-Aldrich
Azure B, certified by the Biological Stain Commission