  • Search Results
  • [Autooxidation of a mixture of lemon essential oils, methyl linolenoate, and methyl oleinate].
[Autooxidation of a mixture of lemon essential oils, methyl linolenoate, and methyl oleinate].
Prikladnaia biokhimiia i mikrobiologiia (2010-11-11)
T A Misharina, M B Terenina, N I Krikunova, I B Medvedeva
ABSTRACT
Stability of components of a mixture of methyl linolenoate and methyl oleinate with two lemon (Citrus limon L.) essential oils in hexane during their autooxidation in light was studied by gas chromatography. The essential oils differed by their quantitative ratio of components: the single-fold (1x) oil contained approximately 90% monoterpene hydrocarbons and 1.47% citral, whereas the proportions of hydrocarbons and citral in the tenfold (10x) oil were approximately 60 and 18.32%, respectively. The concentration and composition of essential oils influence the rates of fatty-acid oxidation and fatty-acid peroxide cleavage. The 1x lemon oil inhibited the oxidation of methyl linolenoate and methyl oleinate, whereas the 10x oil accelerated these processes. The distinctions in the resistance of the major components of lemon essential oil to oxidation, which are determined by their composition and antioxidant properties of unsaturated fatty acids, were revealed.
Sigma-Aldrich
Methyl oleate, 99%
Sigma-Aldrich
Methyl linolenate, ≥99% (GC)
Supelco
Methyl oleate, analytical standard
Supelco
Methyl linolenate, analytical standard
Sigma-Aldrich
Methyl oleate, technical grade, 70%
Sigma-Aldrich
Methyl elaidate, ≥99% (capillary GC)
Supelco
cis-9-Octadecenoic acid methyl ester, certified reference material, 10 mg/mL in heptane
Supelco
Methyl elaidate, analytical standard