Gamma-ray measurements in terrestrial/environmental samples require the use of high efficient detectors because of the low level of the radionuclide activity concentrations in the samples; thus scintillators are suitable for this purpose. Two scintillation detectors were studied in this work; CsI(Tl) and NaI(Tl) with identical size for measurement of terrestrial samples for performance study. This work describes a Monte Carlo method for making the full-energy efficiency calibration curves for both detectors using gamma-ray energies associated with the decay of naturally occurring radionuclides (137)Cs (661keV), (40)K (1460keV), (238)U ((214)Bi, 1764keV) and (232)Th ((208)Tl, 2614keV), which are found in terrestrial samples. The magnitude of the coincidence summing effect occurring for the 2614keV emission of (208)Tl is assessed by simulation. The method provides an efficient tool to make the full-energy efficiency calibration curve for scintillation detectors for any samples geometry and volume in order to determine accurate activity concentrations in terrestrial samples.