  • Search Results
  • A simple well-logging tool using boron-lined sodium iodide scintillators and an 241Am-Be neutron source.
A simple well-logging tool using boron-lined sodium iodide scintillators and an 241Am-Be neutron source.
Radiation protection dosimetry (2012-03-22)
M Rasoulinejad, R Izadi Najafabadi, N Ghal-Eh
ABSTRACT
A couple of 2-inch by 2-inch right cylinder sodium iodide scintillators and an Am-Be radioisotope neutron source have been used in a neutron porosity well-logging tool to explore the variation of hydrogen contents in a prototype formation. Both Monte Carlo N-particle transport code simulation and experimental results of the near- to far-detector responses confirm the reliable sensitivity of proposed tool to the formation porosity.
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, puriss. p.a., ≥99.0% (AT)
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium iodide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium iodide, Vetec, reagent grade