  • Search Results
  • NH4OAc promoted cyclocondensation of 3-(o-allylphenyl)pentane-1,5-diones: synthesis of tetracyclic benzofused 1-azahomoisotwistanes.
NH4OAc promoted cyclocondensation of 3-(o-allylphenyl)pentane-1,5-diones: synthesis of tetracyclic benzofused 1-azahomoisotwistanes.
Organic letters (2012-07-24)
Meng-Yang Chang, Ming-Hao Wu, Hang-Yi Tai
ABSTRACT
A facile two-step synthetic route for preparing the novel tetracyclic skeleton of benzofused 2,6-diaryl-1-azahomoisotwistanes 2 had been developed. The route was carried out by a one-pot tandem cross-coupling reaction of o-allylbenzaldehydes 1 with aryl methyl ketones 3, and NH(4)OAc mediated the cascade cyclocondensation reaction of the resulting 1,5-diketones 4 with the 3-o-allylphenyl group in good yield in two steps.
Product Description
Supelco
Ammonium acetate, suitable for mass spectrometry (MS), LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Pentane, anhydrous, ≥99%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Pentane, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Pentane, reagent grade, 98%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Pentane, ≥99% (GC)
Supelco
Pentane, analytical standard
Sigma-Aldrich
Pentane, spectrophotometric grade, ≥99%
Sigma-Aldrich
Pentane, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Pentane, Laboratory Reagent, ≥95.0% (GC)