  • Search Results
  • Telmisartan inhibits cell proliferation by blocking nuclear translocation of ProHB-EGF C-terminal fragment in colon cancer cells.
Telmisartan inhibits cell proliferation by blocking nuclear translocation of ProHB-EGF C-terminal fragment in colon cancer cells.
PloS one (2013-03-02)
Keiji Ozeki, Satoshi Tanida, Chie Morimoto, Yoshimasa Inoue, Tsutomu Mizoshita, Hironobu Tsukamoto, Takaya Shimura, Hiromi Kataoka, Takeshi Kamiya, Eiji Nishiwaki, Hiroshi Ishiguro, Shigeki Higashiyama, Takashi Joh
ABSTRACT
Current treatment target toward advanced colorectal cancers is mainly focused on the epidermal growth factor receptor (EGFR) signaling, but its additive effects with chemotherapy are still limited. A disintegrin and metalloproteinase (ADAM) cleaves the proheparin-binding epidermal growth factor like growth factor (proHB-EGF). And soluble HB-EGF activates EGFR. In parallel, the carboxy-terminal fragment of proHB-EGF (HB-EGF-CTF) translocates into the inner nuclear membrane, and subsequently exerts on the regulation of cell proliferation by binding nuclear promyelocytic leukemia zinc finger (PLZF) protein, a transcriptional repressor, thereby causing its nuclear export. We hypothesized that the inhibition of HB-EGF-CTF nuclear translocation may be a new strategy in preventing cell proliferation. 12-O-tetradecanoylphorbor-13-acetate (TPA) was treated to activate ADAM. Nine-thousand chemical compounds were screened for their efficacies in blocking the binding of HB-EGF-CTF to promyelocytic leukemia zinc finger (PLZF) with Alphascreen system. The obtained candidates were then used to block the binding of HB-EGF-CTF to PLZF in colon cancer cells, HT29 and HCT116. Cell proliferation was investigated with a growth curve assay. The intracellular localization, and association between HB-EGF-CTF and PLZF, was assessed with immunofluorescent staining, and immunoprecipitation and Western blotting, respectively. The effects of obtained candidates on EGFR phosphorylation and on nuclear translocation of HB-EGF-CTF and export of PLZF during the angiotensin II type1 receptor (AT1R) knockdown were also investigated. Telmisartan and candesartan were found to be potential candidates. Telmisartan inhibited TPA-induced cell proliferation stronger than candesartan. Telmisartan, but not candesartan blocked the nuclear translocation of HB-EGF-CTF, and binding of HB-EGF-CTF to PLZF, during TPA stimulation. Both telmisartan and candesartan did not inhibit TPA-induced EGFR phosphorylation, and telmisartan, but not candesartan, inhibited TPA-induced nuclear translocation of HB-EGF-CTF after knockdown of AT1R. The inhibition of HB-EGF-CTF nuclear translocation with telmisartan may be a novel strategy in preventing cell proliferation.
Product Description
Telmisartan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Telmisartan, ≥98% (HPLC), solid
Telmisartan for system suitability, European Pharmacopoeia (EP) Reference Standard
Telmisartan for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse