  • Search Results
  • TRP and ASIC channels mediate the antinociceptive effect of citronellyl acetate.
TRP and ASIC channels mediate the antinociceptive effect of citronellyl acetate.
Chemico-biological interactions (2013-04-09)
Emiliano Ricardo Vasconcelos Rios, Nayrton Flávio Moura Rocha, Alyne Mara Rodrigues Carvalho, Leonardo Freire Vasconcelos, Marília Leite Dias, Damião Pergentino de Sousa, Francisca Cléa Florenço de Sousa, Marta Maria de França Fonteles
ABSTRACT
Citronellyl acetate (CAT), a monoterpene product of the secondary metabolism of plants, has been shown in the literature to possess several different biological activities. However, no antinociceptive abilities have yet been discussed. Here, we used acute pain animal models to describe the antinociceptive action of CAT. The acetic acid-induced writhing test and the paw-licking test, in which paw licking was induced by glutamate and formalin, were performed to evaluate the antinociceptive action of CAT and to determine the involvement of PKC, PKA, TRPV1, TRPA1, TRPM8 and ASIC in its antinociceptive mechanism. To do so, we induced paw-linking using agonists. CAT was administered intragastrically (25, 50, 75, 100 and 200 mg/kg), and the two higher doses caused antinociceptive effects in the acetic acid model; the highest dose reduced pain for 4h after it was administered (200 mg/kg). In the formalin test, two doses of CAT promoted antinociception in both the early and later phases of the test. The glutamate test showed that its receptors are involved in the antinociceptive mechanism of CAT. Pretreatment with CAT did not alter locomotor activity or motor coordination. In an investigation into the participation of TRP channels and ASICs in CAT's antinociceptive mechanism, we used capsaicin (2.2 μg/paw), cinnamaldehyde (10 mmol/paw), menthol (1.2 mmol/paw) and acidified saline (2% acetic acid, pH 1.98). The results showed that TRPV1, TRPM8 and ASIC, but not TRPA1, are involved in the antinociceptive mechanism. Finally, the involvement of PKC and PKA was also studied, and we showed that both play a role in the antinociceptive mechanism of CAT. The results of this work contribute information regarding the antinociceptive properties of CAT on acute pain and show that, at least in part, TRPV1, TRPM8, ASIC, glutamate receptors, PKC and PKA participate in CAT's antinociceptive mechanism.
Product Description
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Citronellyl acetate, ≥95%, FCC, FG
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of Ph. Eur., BP, 35 wt. %, contains 10% methanol as stabilizer
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Millipore
Bifido Selective Supplement B, suitable for microbiology