Merck
A novel carbon isotope biomarker for dietary sugar.
The Journal of nutrition (2013-04-26)
Dale A Schoeller
ABSTRACT
Light element isotope signatures have been used to study food webs in many ecological niches, including those of ancient humans, but to a far lesser extent in modern humans. A recent paper presented results from a pilot study testing the utility of carbon isotope ratio analysis of alanine to measure an individual's sugar intake in the United States. A strong correlation was found between the enrichment of (13)C in alanine and sugar-sweetened beverage consumption. This candidate biomarker of sugar consumption deserves further consideration as an objective marker for use in the study of the relationship between sugar intake and health.
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥99.0% (NT)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Alanine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Alanine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C