  • Search Results
  • Electrodeposition of nanocrystalline Ni-W coatings with citric acid system.
Electrodeposition of nanocrystalline Ni-W coatings with citric acid system.
Journal of nanoscience and nanotechnology (2013-05-08)
Jingjing Du, Lijian Xu, Baizhen Chen, Li De
ABSTRACT
Ni-W alloy coatings were prepared by electrodeposition technique with citric acid as a complexing agent. The influence of the main technical parameters on W content in the Ni-W alloy coatings were investigated by electron dispersive spectroscopy (EDS), and the microstructure of the coatings was characterized and analyzed by employing X-ray diffractometer (XRD), scanning electron microscope (SEM). The results showed that bright and compact Ni-W alloy coatings with 25.78 wt% W were obtained when the concentration of Na2WO4 x 2H2O was 50 g/L, the concentration of NH4Cl was 20 g/L, the bath pH value was 8.0 and the bath temperature was 70 degrees C. The surface of Ni-W alloy coatings were bright, flat and compact, the mircostructure of the alloy coatings were nanocrystalline and the average grain size was about 38 nm by calculating using Scherrer Equation.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Supelco
Citric acid, analytical standard
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Tungsten, powder, 10 μm, ≥99.99% trace metals basis
Supelco
Citric acid, certified reference material, TraceCERT®
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Tungsten, powder, 12 μm, 99.9% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.127 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.5 mm, ≥99.9% trace metals basis