  • Search Results
  • The influence of surface-activated bonding treatment on the peel strength of Cu-Ni metal clad.
The influence of surface-activated bonding treatment on the peel strength of Cu-Ni metal clad.
Journal of nanoscience and nanotechnology (2013-07-19)
Kyung Hoon Kim, Sung Chul Lim, Hyouk Chon Kwon
ABSTRACT
Three different conditions of surface activation bonding, 0.5 m/min, 1.0 m/min and 1.5 m/min surface activation treatment speed, on copper and nickel substrate were investigated from the view point of adhesion of Cu-Ni clad metal. The specimen treated with surface activation treatment speed 1.0 m/min (Sample B) had higher peel strength properties than 0.5 m/min (Sample A) which has more long plasma treatment time. Observing the interface of clad with TEM and XPS spectra, sample B has relatively thin oxide interface including Cu2O and sample A has thick oxide interface including CuO. From these result, it is suggested that the better adhesion of Cu-Ni clad on the SAB process is not due to the increase of plasma treatment exposure time but due more to the control of forming condition as much as thin Cu2O layer growing epitaxial form.
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Copper(II) oxide on alumina, 14-30 mesh, extent of labeling: 13 wt. % loading
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, bars (random sizes), ≥99.999% trace metals basis
Sigma-Aldrich
Copper, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis