  • Search Results
  • Enhanced hematopoietic protection from radiation by the combination of genistein and captopril.
Enhanced hematopoietic protection from radiation by the combination of genistein and captopril.
International immunopharmacology (2013-01-19)
R M Day, T A Davis, M Barshishat-Kupper, E A McCart, A J Tipton, M R Landauer
ABSTRACT
The hematopoietic system is sensitive to radiation injury, and mortality can occur due to blood cell deficiency and stem cell loss. Genistein and the angiotensin converting enzyme (ACE) inhibitor captopril are two agents shown to protect the hematopoietic system from radiation injury. In this study we examined the combination of genistein with captopril for reduction of radiation-induced mortality from hematopoietic damage and the mechanisms of radiation protection. C57BL/6J mice were exposed to 8.25Gy (60)Co total body irradiation (TBI) to evaluate the effects of genistein and captopril alone and in combination on survival, blood cell recovery, hematopoietic progenitor cell recovery, DNA damage, and erythropoietin production. 8.25Gy TBI resulted in 0% survival after 30days in untreated mice. A single subcutaneous injection of genistein administered 24h before TBI resulted in 72% survival. Administration of captopril in the drinking water, from 1h through 30days postirradiation, increased survival to 55%. Genistein plus captopril increased survival to 95%. Enhanced survival was reflected in a reduction of radiation-induced anemia, improved recovery of nucleated bone marrow cells, splenocytes and circulating red blood cells. The drug combination enhanced early recovery of marrow progenitors: erythroid (CFU-E and BFU-E), and myeloid (CFU-GEMM, CFU-GM and CFU-M). Genistein alone and genistein plus captopril protected hematopoietic progenitor cells from radiation-induced micronuclei, while captopril had no effect. Captopril alone and genistein plus captopril, but not genistein alone, suppressed radiation-induced erythropoietin production. These data suggest that genistein and captopril protect the hematopoietic system from radiation injury via independent mechanisms.
Product Description
Supelco
Captopril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Captopril, meets USP testing specifications
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder