  • Search Results
  • Neuron-specific enolase, nucleotides, nucleosides, purine bases, oxypurines and uric acid concentrations in cerebrospinal fluid of children with meningitis.
Neuron-specific enolase, nucleotides, nucleosides, purine bases, oxypurines and uric acid concentrations in cerebrospinal fluid of children with meningitis.
Brain & development (2003-02-13)
Antonio Rodríguez-Núñez, Elena Cid, Javier Rodríguez-García, Félix Camiña, Santiago Rodríguez-Segade, Manuel Castro-Gago
ABSTRACT
To determine the effects of meningitis on cerebral energy metabolism, cerebrospinal fluid concentrations of adenosine monophosphate, inosine monophosphate, inosine, adenosine, guanosine, adenine, guanine, hypoxanthine, xanthine and urate were determined by high-performance liquid chromatography, and neuron-specific enolase by an enzyme immunoassay method, in 100 children with meningitis (45 bacterial, 46 viral and nine tuberculous), aged between 1 month and 13 years, and in 160 age-matched controls. Compared with controls, patients with bacterial meningitis showed high concentrations of hypoxanthine, xanthine and urate; patients with viral meningitis showed high concentrations of inosine, guanosine, xanthine, urate and neuron-specific enolase; and patients with tuberculous meningitis showed very high concentrations of inosine, xanthine and urate. Xanthine and urate concentrations were significantly higher in patients with tuberculous meningitis than in patients with viral or bacterial meningitis. These results suggest that in the acute stage of bacterial, viral and tuberculous meningitis, neuronal energy metabolism may be altered. The measurement of cerebrospinal xanthine and uric acid concentrations may be useful for the early diagnosis of a tuberculous origin.
Product Description
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Pipecolinic acid, 98%
Sigma-Aldrich
L-Alanine, ≥99.0% (NT)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Supelco
L-Alanine, certified reference material, TraceCERT®
Alanine, European Pharmacopoeia (EP) Reference Standard
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2,3-Pyridinedicarboxylic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Urea, Vetec, reagent grade, 99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Pyruvic acid, 98%
Sigma-Aldrich
Pyruvic acid, 95%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets