  • Search Results
  • [A case of methanol intoxication with optic neuropathy visualized on STIR sequence of MR images].
[A case of methanol intoxication with optic neuropathy visualized on STIR sequence of MR images].
Rinsho shinkeigaku = Clinical neurology (2005-08-27)
Hikaru Nagasawa, Manabu Wada, Shingo Koyama, Toru Kawanami, Keiji Kurita, Takeo Kato
ABSTRACT
We report a 24-year-old man with severe methanol intoxication, who showed cerebral and optic nerve damages on MRI. He was admitted to our hospital with an acute onset of unconsciousness and severe metabolic acidosis, and was immediately treated by hemodialysis. His serum methanol concentration was 261.5 mg/dl. Although this methanol concentration was lethal, he was rescued by intensive care. Three days after admission, brain MRI was performed. Diffusion weighted images showed abnormal hyperintensities in bilateral putamina, subcortical white matter and cerebellar hemispheres. STIR sequence revealed bilateral optic nerve swelling with irregular hyperintense rims. These MRI features might reflect the optic nerve damages, mainly demyelination of the optic nerves caused by a myelinoclastic effect of formic acid, a metabolite of methanol.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.