  • Search Results
  • Variants in the Toll-interacting protein gene are associated with susceptibility to sepsis in the Chinese Han population.
Variants in the Toll-interacting protein gene are associated with susceptibility to sepsis in the Chinese Han population.
Critical care (London, England) (2011-01-12)
Zhenju Song, Jun Yin, Chenling Yao, Zhan Sun, Mian Shao, Yaping Zhang, Zhengang Tao, Peizhi Huang, Chaoyang Tong
ABSTRACT
Deregulated or excessive host immune responses contribute to the pathogenesis of sepsis. Toll-like receptor (TLR) signaling pathways and their negative regulators play a pivotal role in the modulation of host immune responses and the development of sepsis. The objective of this study was to investigate the association of variants in the TLR signaling pathway genes and their negative regulator genes with susceptibility to sepsis in the Chinese Han population. Patients with severe sepsis (n = 378) and healthy control subjects (n = 390) were enrolled. Five genes, namely TLR2, TLR4, TLR9, MyD88 and TOLLIP, were investigated for their association with sepsis susceptibility by a tag single nucleotide polymorphism (SNP) strategy. Twelve tag SNPs were selected based on the data of Chinese Han in Beijing from the HapMap project and genotyped by direct sequencing. The mRNA expression levels of TOLLIP were determined using real-time quantitative Polymerase Chain Reaction (PCR) assays, and concentrations of tumor necrosis factor alpha (TNF-α) and interleukin-6 (IL-6) were measured by enzyme-linked immunosorbent assay (ELISA). Our results showed that the minor C-allele of rs5743867 in TOLLIP was significantly associated with the decreased risk of sepsis (Padj = 0.00062, odds ratio (OR)adj = 0.71, 95% confidence interval (CI) 0.59 to 0.86) after adjustment for covariates in multiple logistic regression analysis. A 3-SNP haplotype block harboring the associated SNP rs5743867 also displayed strong association with omnibus test P value of 0.00049. Haplotype GTC showed a protective role against sepsis (Padj = 0.0012), while haplotype GCT showed an increased risk for sepsis (Padj = 0.00092). After exposure to lipopolysaccharide (LPS), TOLLIP mRNA expression levels in peripheral blood mononuclear cells (PBMCs) from homozygotes for the rs5743867C allele were significantly higher than in heterozygotes and homozygotes for the rs5743867T allele (P = 0.013 and P = 0.01, respectively). Moreover, the concentrations of TNF-α and IL-6 in culture supernatants were significantly lower in the subjects of rs5743867CC genotype than in CT and TT genotype subjects (P = 0.016 and P = 0.003 for TNF-α; P = 0.01 and P = 0.002 for IL-6, respectively). Our findings indicated that the variants in TOLLIP were significantly associated with sepsis susceptibility in the Chinese Han population.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, Ready Made solution, 1 mg/mL
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by trichloroacetic acid extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, FITC conjugate
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, Detoxified