  • Search Results
  • New sulfate-reducing bacteria isolated from Buryatian alkaline brackish lakes: description of Desulfonatronum buryatense sp. nov.
New sulfate-reducing bacteria isolated from Buryatian alkaline brackish lakes: description of Desulfonatronum buryatense sp. nov.
Extremophiles : life under extreme conditions (2013-07-25)
Yana Ryzhmanova, Yana Nepomnyashchaya, Tatyana Abashina, Elena Ariskina, Olga Troshina, Mikhail Vainshtein, Victoria Shcherbakova
ABSTRACT
New strains of sulfate-reducing bacteria were isolated from two alkaline brackish lakes located in the Siberian region of Russia, namely in the Southern Transbaikalia, Buriatia. The article presents data describing morphology, physiology, and biochemical characteristics of the isolated strains. These strains Ki4, Ki5, and Su2 were mesophilic and alkaliphilic with optimal growth at pH 8.9, 9.4, and 10.0, respectively. All isolated strains utilized lactate, formate, and ethanol in the presence of sulfate for growth and sulfidogenesis accompanied with formation of acetate and CO2. Strains Ki5 and Su2 were able to reduce Fe(III). The DNA G + C content in strains Ki4, Ki5 and Su2 was 56.3, 48.8 and 59.6 mol%, respectively. According to phylogenetic analysis of 16S rRNA sequences, the new strains were clustered within the genus Desulfonatronum, and the closest relative D. lacustre Z-7951(T) (=DSM 10312(T)) showed 99.3-99.6 % similarity. DNA-DNA relatedness values of the strains Ki4, Ki5, and Su2 with D. lacustre Z-7951(T) were 89, 53, and 79 %, respectively. Polyphasic taxonomy data suggest that strain Ki5(T) is representative of the proposed novel species Desulfonatronum buryatense sp. nov.
Product Description
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Potassium formate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Sigma-Aldrich
Cesium formate, 98%
Supelco
Calcium formate, Standard for quantitative NMR, TraceCERT®
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Calcium formate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium formate-13C, 99 atom % 13C
Sigma-Aldrich
Potassium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Thallium(I) formate, 97%
Sigma-Aldrich
Basosiv M050, produced by BASF