  • Search Results
  • PUA/PSS multilayer coated CaCO3 microparticles as smart drug delivery vehicles.
PUA/PSS multilayer coated CaCO3 microparticles as smart drug delivery vehicles.
Materials science & engineering. C, Materials for biological applications (2013-08-06)
Chao Du, Jun Shi, Jin Shi, Li Zhang, Shaokui Cao
ABSTRACT
Hybrid CaCO3 microparticles coated by sodium poly(styrene sulfonate) (PSS) and aliphatic poly(urethane-amine) (PUA) were developed as thermal-/pH-responsive drug delivery vehicles via LbL self-assembly technique. The DOX release from the CaCO3 microparticles was higher than 60% within 36 h, whereas the value of PUA/PSS-coated microparticles was only 20%. The results demonstrated that the PUA/PSS multilayer coating could reduce the drug release rate and significantly assuage the initial burst release of DOX. In addition, the drug release of the hybrid microparticles was found to be thermal-/pH-dual responsive. More interestingly, more than 90% of DOX was released in 36 h at pH2.1 and 55 °C owing to the combined action of the dissolution of the CaCO3 core and the shrinkage of aliphatic PUA.
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Supelco
Calcium carbonate, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, Vetec, reagent grade, 99%
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ReagentPlus®