  • Search Results
  • Trophic dynamics of U, Ni, Hg and other contaminants of potential concern on the Department of Energy's Savannah River Site.
Trophic dynamics of U, Ni, Hg and other contaminants of potential concern on the Department of Energy's Savannah River Site.
Environmental monitoring and assessment (2013-08-28)
Paul G Edwards, Karen F Gaines, A Lawrence Bryan, James M Novak, Susan A Blas
ABSTRACT
The Department of Energy's Savannah River Site is a former nuclear weapon material production and current research facility located in South Carolina, USA. Wastewater discharges from a fuel and nuclear reactor target manufacturing facility released depleted and natural U, as well as other metals into the Tims Branch-Steed Pond water system. We investigated the current dynamics of this system for the purposes of environmental monitoring and assessment by examining metal concentrations, bioavailability, and trophic transfer of contaminants in seven ponds. Biofilm, detritus, and Anuran and Anisopteran larvae were collected and analyzed for stable isotopes (δ (15)N, δ (13)C) and contaminants of potential concern (COPC) with a focus on Ni, U, and Hg, to examine metal mobility. Highest levels of Ni and U were found in biofilms U (147 and 332 mg kg(-1) DW, respectively), while highest Hg levels were found in tadpoles (1.1 mg kg(-1) DW). We found intraspecific biomagnification of COPCs as expressed through stable isotope analysis. Biofilms were the best indicators for contamination and Anuran larvae with the digestive tract removed were the best indicators of the specific bioavailability of the focal metals. Monitoring data showed that baseline δ (15)N values differed between ponds, but within a pond, values were stable throughout tadpole Gosner stage, strengthening the case to use this species for monitoring purposes. It is likely that there still is risk to ecosystem integrity as COPC metals are being assimilated into lower trophic organisms and even low levels of this mixture has shown to produce deleterious effects to some wildlife species.
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis