  • Search Results
  • Calcium carbonate does not affect imatinib pharmacokinetics in healthy volunteers.
Calcium carbonate does not affect imatinib pharmacokinetics in healthy volunteers.
Cancer chemotherapy and pharmacology (2013-10-31)
Hussein Tawbi, Susan M Christner, Yan Lin, Matthew Johnson, Emily T Mowrey, Craig Cherrin, Edward Chu, James J Lee, Shannon Puhalla, Ronald Stoller, Leonard R Appleman, Brian M Miller, Jan H Beumer
ABSTRACT
Imatinib mesylate (Gleevec(®)/Glivec(®)) has revolutionized the treatment of chronic myeloid leukemias and gastrointestinal stromal tumors, and there is evidence for an exposure response relationship. Calcium carbonate is increasingly used as a calcium supplement and in the setting of gastric upset associated with imatinib therapy. Calcium carbonate could conceivably elevate gastric pH and complex imatinib, thereby influencing imatinib absorption and exposure. We aimed to evaluate whether use of calcium carbonate has a significant effect on imatinib pharmacokinetics. Eleven healthy subjects were enrolled in a 2-period, open-label, single-institution, randomized crossover, fixed-schedule study. In one period, each subject received 400 mg of imatinib p.o. In the other period, 4,000 mg calcium carbonate (Tums Ultra(®)) was administered p.o. 15 min before 400 mg of imatinib. Plasma concentrations of imatinib and its active N-desmethyl metabolite CGP74588 were assayed by LC-MS; data were analyzed non-compartmentally and compared after log transformation. Calcium carbonate administration did not significantly affect the imatinib area under the plasma concentration versus time curve (AUC) (41.2 μg/mL h alone vs. 40.8 μg/mL h with calcium carbonate, P = 0.99), maximum plasma concentration (C(max)) (2.35 μg/mL alone vs. 2.39 μg/mL with calcium carbonate, P = 0.89). Our results indicate that the use of calcium carbonate does not significantly affect imatinib pharmacokinetics.
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, Vetec, reagent grade, 99%
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Supelco
Calcium carbonate, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis