Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis