  • Search Results
  • Aroma profiles and preferences of Jasminum sambac L. flowers grown in Thailand.
Aroma profiles and preferences of Jasminum sambac L. flowers grown in Thailand.
Journal of cosmetic science (2014-01-09)
Mayuree Kanlayavattanakul, Sarun Kitsiripaisarn, Nattaya Lourith
ABSTRACT
Comparison of volatile constituents and odor preference of Jasminum sambac cultivated in Thailand was performed by enfleurage and solvent extractions. Enfleurage bases consisting of spermaceti wax, olive, sunflower, and rice bran oils were prepared. The defleurage flower was daily replaced with fresh jasmine for a period of 12 days. The absolute de pomades and extraits of each base were subjected to gas chromatography mass spectrometry (GC/MS) analysis, comparing with the concrete and absolute values obtained from maceration of jasmine in n-hexane for 24 h. Linalool, benzyl acetate, and α-farnesene were found as the main volatile compounds in the jasmine extracts. Spermaceti wax and olive oil gave the best quality base, exhibiting the most preferred resemblance of jasmine odor with the least difference from fresh jasmine, as evaluated by 103 Thai volunteers.
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Linalool, 97%
Supelco
Hexane, analytical standard
Supelco
Linalool, analytical standard
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Benzyl acetate, ≥99%
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Sigma-Aldrich
Benzyl acetate, ≥99%, FCC, FG
Supelco
Benzyl acetate, analytical standard
Supelco
(−)-Linalool, analytical standard
Sigma-Aldrich
Benzyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Hexane-1-13C, 99 atom % 13C