STED microscopy with continuous wave beams.
Nature methods (2007-10-24)
Katrin I Willig, Benjamin Harke, Rebecca Medda, Stefan W Hell
ABSTRACT
We report stimulated emission depletion (STED) fluorescence microscopy with continuous wave (CW) laser beams. Lateral fluorescence confinement from the scanning focal spot delivered a resolution of 29-60 nm in the focal plane, corresponding to a 5-8-fold improvement over the diffraction barrier. Axial spot confinement increased the axial resolution by 3.5-fold. We observed three-dimensional (3D) subdiffraction resolution in 3D image stacks. Viable for fluorophores with low triplet yield, the use of CW light sources greatly simplifies the implementation of this concept of far-field fluorescence nanoscopy.
Sigma-Aldrich
Atto 647N NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Phalloidin–Atto 647N, BioReagent, suitable for fluorescence, ≥80% (HPLC)
Supelco
Atto 565 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Anti-Rabbit-IgG - Atto 647N antibody produced in goat, 1 mg/mL IgG
Sigma-Aldrich
Anti-Mouse-IgG - Atto 647N antibody produced in goat, contains 50% glycerol as stabilizer
Sigma-Aldrich
Atto 532 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Atto 647N, BioReagent, suitable for fluorescence, ≥98.0% (HPCE)
Supelco
Atto 647N maleimide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
SAFC
Atto 465 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
NTA - Atto 647 N, BioReagent, suitable for fluorescence, ≥80% (HPCE)
Sigma-Aldrich
Atto 647N-Streptavidin, BioReagent, suitable for fluorescence
Supelco
Atto 590 NHS ester, BioReagent, suitable for fluorescence, ≥60% (coupling to amines)
Sigma-Aldrich
Atto 425 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 565, BioReagent, suitable for fluorescence, ≥75% (sum of isomers, HPCE/HPLC)
Sigma-Aldrich
Atto 647N Protein Labeling Kit
Sigma-Aldrich
Atto 532, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 590-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPCE)
Sigma-Aldrich
Atto 647N-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Atto 590, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 565-Biotin, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Phalloidin–Atto 390, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Atto 390 NHS ester, BioReagent, suitable for fluorescence
Sigma-Aldrich
Phalloidin–Atto 532, suitable for fluorescence, ≥95.0% (HPCE)
Sigma-Aldrich
Atto 590-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N amine
Sigma-Aldrich
Phalloidin–Atto 425, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647N azide
Sigma-Aldrich
Phalloidin–Atto 590, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 425 maleimide, BioReagent, suitable for fluorescence, ≥70% (coupling to thiols)