  • Search Results
  • Birefringent device converts a standard scanning microscope into a STED microscope that also maps molecular orientation.
Birefringent device converts a standard scanning microscope into a STED microscope that also maps molecular orientation.
Optics express (2010-02-23)
Matthias Reuss, Johann Engelhardt, Stefan W Hell
ABSTRACT
Stimulated emission depletion (STED) microscopy usually employs a scanning excitation beam that is superimposed by a donut-shaped STED beam for keeping the fluorophores at the periphery of the excitation spot dark. Here, we introduce a simple birefringent device that produces a donut-shaped focal spot with suitable polarization for STED, while leaving the excitation spot virtually intact. The device instantly converts a scanning (confocal) microscope with a co-aligned STED beam into a full-blown STED microscope. The donut can be adapted to reveal, through the resulting fluorescence image, the orientation of fluorophores in the sample, thus directly providing subdiffraction resolution images of molecular orientation.
Sigma-Aldrich
Atto 647N NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Phalloidin–Atto 647N, BioReagent, suitable for fluorescence, ≥80% (HPLC)
Supelco
Atto 565 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Anti-Rabbit-IgG - Atto 647N antibody produced in goat, 1 mg/mL IgG
Sigma-Aldrich
Anti-Mouse-IgG - Atto 647N antibody produced in goat, contains 50% glycerol as stabilizer
Sigma-Aldrich
Atto 532 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Atto 647N, BioReagent, suitable for fluorescence, ≥98.0% (HPCE)
Supelco
Atto 647N maleimide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
SAFC
Atto 465 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
NTA - Atto 647 N, BioReagent, suitable for fluorescence, ≥80% (HPCE)
Sigma-Aldrich
Atto 647N-Streptavidin, BioReagent, suitable for fluorescence
Supelco
Atto 590 NHS ester, BioReagent, suitable for fluorescence, ≥60% (coupling to amines)
Sigma-Aldrich
Atto 425 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 565, BioReagent, suitable for fluorescence, ≥75% (sum of isomers, HPCE/HPLC)
Sigma-Aldrich
Atto 647N Protein Labeling Kit
Sigma-Aldrich
Atto 532, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 590-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPCE)
Sigma-Aldrich
Atto 647N-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Atto 590, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 565-Biotin, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Phalloidin–Atto 390, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Atto 390 NHS ester, BioReagent, suitable for fluorescence
Sigma-Aldrich
Phalloidin–Atto 532, suitable for fluorescence, ≥95.0% (HPCE)
Sigma-Aldrich
Atto 590-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N amine
Sigma-Aldrich
Phalloidin–Atto 425, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647N azide
Sigma-Aldrich
Phalloidin–Atto 590, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 425 maleimide, BioReagent, suitable for fluorescence, ≥70% (coupling to thiols)