  • Search Results
  • CT angiography of the neck: value of contrast medium dose reduction with low tube voltage and high tube current in a 64-detector row CT.
CT angiography of the neck: value of contrast medium dose reduction with low tube voltage and high tube current in a 64-detector row CT.
Clinical radiology (2014-02-08)
W Xia, J-T Wu, X-R Yin, Z-J Wang, H-T Wu
ABSTRACT
To evaluate the feasibility of a low-dose contrast medium protocol for 64-detector row computed tomography angiography (CTA) of the neck using a low-tube-voltage/high-tube-current setting. A phantom study was performed using 64-detector row spiral CT at multiple tube voltage and current settings. Iodine contrast medium attenuation curves were acquired by processing and used to select the best contrast medium-to-noise ratio (CNR). A prospective clinical study was then performed on 84 patients requiring neck CTA. Patients were randomly divided into two groups of 42. Group A was examined using the conventional imaging protocol (120 kV, 400 mAs) and group B was examined at 80 kV and 600 mAs along with a 50% reduction in contrast medium dose. The CT dose index-volume (CTDI(vol)), background noise (BN), and CNR were measured and statistically analysed. Various image quality criteria were evaluated by two senior radiologists using a qualitative five-point scale. Comparing group B with A, CTDIvol decreased by 54% (B: 27.48 mGy, A: 59.11 mGy), however, the CNR increased by 50%. The mean attenuation, which was caused by venous streak artefacts, was significantly lower in group B than A. Qualitative image analysis found that all criteria were significantly better for group B than A. At 64-detector row spiral CT, the low-tube-voltage/high-tube-current with low-dose contrast medium protocol was superior to the conventional protocol regarding radiation dose, venous streak artefacts, and image quality, and is feasible for CTA of the neck.
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, Vetec, reagent grade, 99%
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)