  • Search Results
  • Analysis of perfusates during hypothermic machine perfusion by NMR spectroscopy: a potential tool for predicting kidney graft outcome.
Analysis of perfusates during hypothermic machine perfusion by NMR spectroscopy: a potential tool for predicting kidney graft outcome.
Transplantation (2014-04-01)
Delphine Bon, Claire Billault, Billault Claire, Raphaël Thuillier, William Hebrard, Nadège Boildieu, Olivier Celhay, Jacques Irani, François Seguin, Thierry Hauet
ABSTRACT
Machine perfusion use has been reported to promote graft outcome in case of donation after cardiac death. Our objective was to evaluate the potential for nuclear magnetic resonance (NMR) to predict graft outcome by analyzing perfusates during machine perfusion time. We used a renal autotransplantation model mimicking deceased after cardiac death donors with pigs. Organs were subjected to 60 min of warm ischemia before the hypothermic machine preservation during 22 hr. We studied the correlation between creatinemia after transplantation and the NMR data from perfusates. A metabonomic analysis allowed us to highlight the evolution of several metabolites during perfusion: the concentration of lactate, choline, or amino acids such as valine, glycine, or glutamate increased with time, whereas there was a diminution of total glutathione during this period. The changes in these biomarkers were less severe in the group with the better outcome. Statistical analysis revealed a strong association between the level of those metabolites during machine perfusion and function recovery (Spearman rank ≥0.89; P<0.05). Multivariate analysis of lesion biomarkers during kidney perfusion using NMR data could be an interesting tool to assess graft quality, particularly because analyses times (2 hr total) are compatible with clinical application.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
L-Glutathione reduced, BioReagent, suitable for cell culture, ≥98.0%, powder
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Lactic acid, 85%, FCC
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Lactic acid, natural, ≥85%
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
USP
Lactic acid, United States Pharmacopeia (USP) Reference Standard
USP
Choline chloride, United States Pharmacopeia (USP) Reference Standard