  • Search Results
  • Epinephrine stimulation of anion secretion in the Calu-3 serous cell model.
Epinephrine stimulation of anion secretion in the Calu-3 serous cell model.
American journal of physiology. Lung cellular and molecular physiology (2014-04-08)
Amiraj Banga, Stephanie Flaig, Shanta Lewis, Seth Winfree, Bonnie L Blazer-Yost
ABSTRACT
Calu-3 is a well-differentiated human bronchial cell line with the characteristics of the serous cells of airway submucosal glands. The submucosal glands play a major role in mucociliary clearance because they secrete electrolytes that facilitate airway hydration. Given the significance of both long- and short-term β-adrenergic receptor agonists in the treatment of respiratory diseases, it is important to determine the role of these receptors and their ligands in normal physiological function. The present studies were designed to characterize the effect of epinephrine, the naturally occurring β-adrenergic receptor agonist, on electrolyte transport of the airway serous cells. Interestingly, epinephrine stimulated two anion secretory channels, the cystic fibrosis transmembrane conductance regulator and a Ca(2+)-activated Cl(-) channel, with the characteristics of transmembrane protein 16A, thereby potentially altering mucociliary clearance via multiple channels. Consistent with the dual channel activation, epinephrine treatment resulted in increases in both intracellular cAMP and Ca(2+). Furthermore, the present results extend previous reports indicating that the two anion channels are functionally linked.
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Adrenaline, European Pharmacopoeia (EP) Reference Standard
Adrenaline with impurity F, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
(−)-Epinephrine
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)
USP
Amiloride hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ionomycin from Streptomyces conglobatus, ≥98% (HPLC)