Addendum to Fragrance material review on linalool.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-12-20)
A Lapczynski, C S Letizia, A M Api
ABSTRACT
A Fragrance material review on linalool was published by Letizia et al. in Food and Chemical Toxicology 41 (2003) 943-964. This addendum to the earlier publication will only report studies that were conducted after the Fragrance material review was published.
Sigma-Aldrich
Linalool, 97%
Supelco
Linalool, analytical standard
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Supelco
(−)-Linalool, analytical standard