Recent advances in the biological production of mannitol.
Applied microbiology and biotechnology (2009-07-07)
Seung Hoon Song, Claire Vieille
ABSTRACT
Mannitol is a fructose-derived, 6-carbon sugar alcohol that is widely found in bacteria, yeasts, fungi, and plants. Because of its desirable properties, mannitol has many applications in pharmaceutical products, in the food industry, and in medicine. The current mannitol chemical manufacturing process yields crystalline mannitol in yields below 20 mol% from 50% glucose/50% fructose syrups. Thus, microbial and enzymatic mannitol manufacturing methods have been actively investigated, in particular in the last 10 years. This review summarizes the most recent advances in biological mannitol production, including the development of bacterial-, yeast-, and enzyme-based transformations.
Sigma-Aldrich
D-Mannitol, ≥98%
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Mannitol, European Pharmacopoeia (EP) Reference Standard
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination