  • Search Results
  • Various novel erythromycin derivatives obtained by different modifications: recent advance in macrolide antibiotics.
Various novel erythromycin derivatives obtained by different modifications: recent advance in macrolide antibiotics.
Mini reviews in medicinal chemistry (2010-05-18)
C Ma, S Ma
ABSTRACT
The rapid emergence of drug resistance intensified the search for new antimicrobial agents, leading to lots of novel derivatives obtained from 14- and 15-membered macrolides by chemical modifications. Many of them exhibited enhanced antibacterial activity and expanded antibacterial spectrum. Especially some of them were found to be potent for the treatment of multi-drug-resistant bacterial infections. Besides, the other biological effects of macrolide derivatives were also found. In this article, we reviewed the recent advance in the novel macrolide derivatives designed by different structural modifications on erythronolide skeleton, cladinose and desosamine in the structures.
Product Description
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard