Merck
Telmisartan for the management of patients at high cardiovascular risk.
Current medical research and opinion (2011-07-02)
Luis M Ruilope
ABSTRACT
Cardiovascular disease (CVD) places a significant burden on healthcare providers. High blood pressure (BP) is the single most prevalent risk factor for CVD worldwide and is responsible for more deaths than any other risk factor. 'Cardiovascular (CV) high-risk patients' make up the broad cross-section of patients in the middle of the risk spectrum for CVD progression that is referred to as the CV continuum and includes those with atherothrombotic disease, those with target organ damage associated with type 2 diabetes and those with multiple risk factors. Angiotensin II is involved in CVD progression at every stage of the CV continuum, making the renin-angiotensin system a rational target for pharmacologic intervention. Angiotensin II receptor blockers (ARBs) offer a better tolerated alternative to angiotensin converting enzyme inhibitors, with greater long-term adherence. The ARB telmisartan recently received an indication for CV prevention. A PubMed literature search was conducted to identify evidence on the use of telmisartan for preventing CV events. Telmisartan has a favourable safety and tolerability profile, and has demonstrated efficacious and long-lasting 24-hour BP reductions, whether as monotherapy or in combination with hydrochlorothiazide or amlodipine. In the largest CV prevention trial program undertaken with an ARB (the ONgoing Telmisartan Alone and in combination with Ramipril Global Endpoint Trial; ONTARGET), telmisartan 80 mg/day alone was as effective as ramipril in reducing the composite primary endpoint of CV mortality, non-fatal myocardial infarction, non-fatal stroke and hospitalization for heart failure in CV high-risk patients. However, patients were significantly more likely to adhere to treatment with telmisartan than ramipril due to its better tolerability. To date, telmisartan is the only ARB indicated to reduce CV morbidity in a broad CV high-risk population.
Product Description
Telmisartan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Telmisartan, ≥98% (HPLC), solid
Telmisartan for system suitability, European Pharmacopoeia (EP) Reference Standard
Telmisartan for peak identification, European Pharmacopoeia (EP) Reference Standard