  • Search Results
  • Arachidonic acid and lipoxin A4 as possible endogenous anti-diabetic molecules.
Arachidonic acid and lipoxin A4 as possible endogenous anti-diabetic molecules.
Prostaglandins, leukotrienes, and essential fatty acids (2013-01-09)
Undurti N Das
ABSTRACT
In both type 1 and type 2 diabetes mellitus, increased production of pro-inflammatory cytokines and reactive oxygen species (ROS) occurs that induce apoptosis of β cells and cause peripheral insulin resistance respectively though the degree of their increased production is higher in type 1 and less in type 2 diabetes mellitus. Despite this, the exact mechanism(s) that lead to increased production of pro-inflammatory cytokines: interleukin-6 (IL-6) and tumor necrosis factor-α (TNF-α) and ROS is not known. Studies showed that plasma concentrations of arachidonic acid (AA) and lipoxin A4 (LXA4) are low in alloxan-induced type 1 diabetes mellitus in experimental animals and patients with type 2 diabetes mellitus. Prior administration of AA, eicosapentaenoic and docosahexaenoic acids (EPA and DHA, respectively) and transgenic animals that produce increased amounts of EPA and DHA acids were protected from chemical-induced diabetes mellitus that was associated with enhanced formation of LXA4 and resolvins, while protectin D1 ameliorated peripheral insulin resistance. AA, LXA4, resolvins and protectins inhibit IL-6 and TNF-α production and suppress ROS generation. Thus, AA and lipoxins, resolvins and protectins may function as endogenous anti-diabetic molecules implying that their administration could be useful in the prevention and management of both types of diabetes mellitus.
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)