  • Search Results
  • Effect of folic acid, vitamin B6 and vitamin B12 supplementation on mortality and cardiovascular complication among patients with chronic kidney disease: an evidence-based case report.
Effect of folic acid, vitamin B6 and vitamin B12 supplementation on mortality and cardiovascular complication among patients with chronic kidney disease: an evidence-based case report.
Acta medica Indonesiana (2013-06-19)
Alvin Nursalim, Parlindungan Siregar, Indah S Widyahening
ABSTRACT
to determine whether the administration of folic acid, vitamin B6 and vitamin B12 would lead to reduction of cardiovascular complication and mortality among CKD patients. a search was conducted on PubMed and Google. The selection of title and abstract was conducted using inclusion and exclusion criterias, which led to six relevant articles. The selected studies were critically appraised for its validity, importance and applicability. the administration of folic acid and vitamin B reduce homocysteine level among CKD patients. Despite homocysteine level reduction, all six studies reported similar findings that folic acid and vitamin B supplementation did not significantly reduce cardiovascular complication and mortality. treatment with folic acid, vitamin B6 and vitamin B12 did not reduce cardiovascular complication and mortality among CKD patients.
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Folic acid, meets USP testing specifications
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cyanocobalamin (B12), analytical standard
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Folic acid, Vetec, reagent grade, ≥97%
USP
Folic acid, United States Pharmacopeia (USP) Reference Standard
Folic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, Vetec, reagent grade, ≥98%