Heme-dependent dioxygenases in tryptophan oxidation.
Archives of biochemistry and biophysics (2013-12-04)
Jiafeng Geng, Aimin Liu
ABSTRACT
L-Tryptophan is an essential amino acid for mammals. It is utilized not only for protein synthesis but also for the biosynthesis of serotonin and melatonin by the serotonin pathway as well as nicotinamide adenine dinucleotide by the kynurenine pathway. Although the kynurenine pathway is responsible for the catabolism of over 90% of l-tryptophan in the mammalian intracellular and extracellular pools, the scientific field was dominated in the last century by studies of the serotonin pathway, due to the physiological significance of the latter's catabolic intermediates and products. However, in the past decade, the focus gradually reversed as the link between the kynurenine pathway and various neurodegenerative disorders and immune diseases is increasingly highlighted. Notably, the first step of this pathway, which is catalyzed by heme-dependent dioxygenases, has been proven to be a potential target for immune regulation and cancer treatment. A thorough understanding of the intriguing chemistry of the heme-dependent dioxygenases may yield insight for the drug discovery of these prevalent illnesses. In this review, we survey enzymatic and mechanistic studies, initially started by Kotake and Masayama over 70 years ago, at the molecular level on the heme-dependent tryptophan dioxygenation reactions.
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Tryptophan, certified reference material, TraceCERT®
Tryptophan, European Pharmacopoeia (EP) Reference Standard