Merck
Sodium and potassium in health and disease.
Metal ions in life sciences (2014-01-29)
Hana R Pohl, John S Wheeler, H Edward Murray
ABSTRACT
Sodium and potassium are essential for human health. They are important ions in the body and are associated with many physiologic and pathophysiologic processes. The chapter summarizes the basic physiologic actions of sodium and potassium on membranes of the neurologic and muscular systems. It provides information regarding the kinetics, i.e., absorption, distribution, and excretion of these ions and their movement between the intracellular and extracellular compartments. It also explains the physiologic systems that can influence proper homeostasis between sodium and potassium. Concentrations of sodium in the blood that exceed or do not reach the normal value range are called hypernatremia or hyponatremia, respectively. Similarly, the clinicians recognize hyperkalemia and hypokalemia. Pathologies associated with these states are described and examples of some of the diseases are presented here.
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis