  • Search Results
  • Use of a trypsin, Peru balsam, and castor oil spray on the oral mucosa: case report and review of the literature.
Use of a trypsin, Peru balsam, and castor oil spray on the oral mucosa: case report and review of the literature.
Pharmacotherapy (1989-01-01)
T A Noble, D S Carr, M F Gonzalez
ABSTRACT
Granulex is an aerosolized spray (Dow B. Hickman Pharmaceuticals, Sugar Land, TX 77487) that contains trypsin, Peru balsam, and castor oil. It has been available for many years as a topical spray for the treatment of decubitus ulcers. We used Granulex to promote tissue healing of a necrotic ulcer of the oral mucosa in a patient with advanced oropharyngeal squamous cell carcinoma.
Sigma-Aldrich
Castor oil
Sigma-Aldrich
Castor oil, meets testing specifications per USP
Sigma-Aldrich
Castor oil, tested according to Ph. Eur.