  • Search Results
  • Norfloxacin in the treatment of gonorrhea due to penicillinase and non-penicillinase producing Neisseria gonorrheae: a review.
Norfloxacin in the treatment of gonorrhea due to penicillinase and non-penicillinase producing Neisseria gonorrheae: a review.
Scandinavian journal of infectious diseases. Supplementum (1988-01-01)
C T Lee, E C Wong
ABSTRACT
In 12 recent clinical studies, norfloxacin given in various dosage regimens was shown to be highly effective in treating 1,588 gonococcal infections in 1,486 patients. A single oral dose of norfloxacin 800 mg cured 99.2% of 783 patients with penicillinase producing strains of Neisseria gonorrheae (PPNG) and non-penicillinase producing N. gonorrheae (NPPNG) causing urethritis, cervicitis and anorectal infections. A single oral dose of 800 mg of norfloxacin was found to be an effective alternative to a 2 g intramuscular dose of spectinomycin and superior to a 2.5 g oral dose of thiamphenicol for the treatment of PPNG urethritis or cervicitis. Its efficacy in treatment of NPPNG infections was shown to be similar to that of a single oral dose of 3.5 g of ampicillin plus 1 g of probenecid. Mild and transient gastrointestinal and neurological adverse events occurred in 3.3% of patients treated with norfloxacin 800 mg. Norfloxacin 800 mg as a single dose is recommended as an effective and safe single-dose treatment of urethritis, cervicitis and anorectal infections caused by PPNG or NPPNG.
Product Description
Norfloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Norfloxacin, European Pharmacopoeia (EP) Reference Standard
Norfloxacin for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Norfloxacin, VETRANAL®, analytical standard