  • Search Results
  • Metabolism of dietary stearic acid relative to other fatty acids in human subjects.
Metabolism of dietary stearic acid relative to other fatty acids in human subjects.
The American journal of clinical nutrition (1994-12-01)
E A Emken
ABSTRACT
This paper reviews results obtained by stable-isotope-tracer methods for stearic acid (18:0) and palmitic acid (16:0) metabolism and the influence of dietary linoleic acid on the metabolism of these saturated fatty acids in humans subjects. The results, based on stable-isotope-tracer data, show that absorption of 18:0 is not significantly different from 16:0; percent desaturation of 18:0 to 9-cis 18:1 (9.2%) is 2.4 times higher than for 16:0 to 9-cis 16:1 (3.9%) and 9-desaturation is not greatly influenced by the amount of linoleic acid in typical US diets. Additionally, compared with 16:0, 18:0 incorporation is 30-40% lower for plasma triglyceride and cholesterol ester and approximately 40% higher for phosphatidylcholine; beta-oxidation of saturated fatty acids was slower than for unsaturated fatty acids and increasing the intake of dietary linoleic acid decreased beta-oxidation of saturated fatty acids. These results indicate that metabolic differences between 18:0 and 16:0 only partially explain the difference in the cholesterolemic effect reported for these saturated fatty acids.
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, analytical standard
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Stearic acid, Vetec, reagent grade, 94%