  • Search Results
  • Sustained delivery of rhBMP-2 by means of poly(lactic-co-glycolic acid) microspheres: cranial bone regeneration without heterotopic ossification or craniosynostosis.
Sustained delivery of rhBMP-2 by means of poly(lactic-co-glycolic acid) microspheres: cranial bone regeneration without heterotopic ossification or craniosynostosis.
Plastic and reconstructive surgery (2014-03-14)
Jason D Wink, Patrick A Gerety, Rami D Sherif, Youngshin Lim, Nadya A Clarke, Chamith S Rajapakse, Hyun-Duck Nah, Jesse A Taylor
ABSTRACT
Commercially available recombinant human bone morphogenetic protein 2 (rhBMP2) has demonstrated efficacy in bone regeneration, but not without significant side effects. The authors used rhBMP2 encapsulated in poly(lactic-co-glycolic acid) (PLGA) microspheres placed in a rabbit cranial defect model to test whether low-dose, sustained delivery can effectively induce bone regeneration. The rhBMP2 was encapsulated in 15% PLGA using a double-emulsion, solvent extraction/evaporation technique, and its release kinetics and bioactivity were tested. Two critical-size defects (10 mm) were created in the calvaria of New Zealand white rabbits (5 to 7 months of age, male and female) and filled with a collagen scaffold containing either (1) no implant, (2) collagen scaffold only, (3) PLGA-rhBMP2 (0.1 μg per implant), or (4) free rhBMP2 (0.1 μg per implant). After 6 weeks, the rabbits were killed and defects were analyzed by micro-computed tomography, histology, and finite element analysis. The rhBMP2 delivered by means of bioactive PLGA microspheres resulted in higher volumes and surface area coverage of new bone than an equal dose of free rhBMP2 by micro-computed tomography (p=0.025 and p=0.025). Finite element analysis indicated that the mechanical competence using the regional elastic modulus did not differ with rhBMP2 exposure (p=0.70). PLGA-rhBMP2 did not demonstrate heterotopic ossification, craniosynostosis, or seroma formation. Sustained delivery by means of PLGA microspheres can significantly reduce the rhBMP2 dose required for de novo bone formation. Optimization of the delivery system may be a key to reducing the risk for recently reported rhBMP2-related adverse effects.
Product Description
USP
Lactic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Lactic acid, natural, ≥85%
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of Ph. Eur., BP, 35 wt. %, contains 10% methanol as stabilizer
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard
Millipore
Dichloromethane, suitable for HPLC, ≥99.8% (GC)
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Supelco
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactic acid solution, Vetec, reagent grade, 85%