  • Search Results
  • The thyroxine-containing thyroglobulin peptide (aa 2549-2560) is a target epitope in iodide-accelerated spontaneous autoimmune thyroiditis.
The thyroxine-containing thyroglobulin peptide (aa 2549-2560) is a target epitope in iodide-accelerated spontaneous autoimmune thyroiditis.
Journal of immunology (Baltimore, Md. : 1950) (2014-06-01)
Panayota Kolypetri, Karen Carayanniotis, Shofiur Rahman, Paris E Georghiou, Vassiliki Magafa, Paul Cordopatis, George Carayanniotis
ABSTRACT
Enhanced iodide ingestion is known to accelerate the incidence and severity of spontaneous autoimmune thyroiditis [iodide-accelerated spontaneous autoimmune thyroiditis (ISAT)] in NOD.H2(h4) mice. CD4+ cells are required for the development and maintenance of ISAT, but their target epitopes remain unknown. In this study, we show that the previously identified thyroglobulin (Tg) T cell epitope p2549-2560 containing thyroxine at position 2553 (T4p2553) induces thyroiditis as well as strong specific T and B cell responses in NOD.H2(h4) mice. In ISAT, activated CD4+ T cells specific for T4p2553 are detected before the disease onset in thyroid-draining cervical lymph nodes only in mice placed on an iodide-rich diet and not in age-matched controls. In addition, selective enrichment of CD4+ IFN-γ+ T4p2553-specific cells is observed among cervical lymph node cells and intrathyroidal lymphocytes. T4p2553 was equally detectable on dendritic cells obtained ex vivo from cervical lymph node cells of NaI-fed or control mice, suggesting that the iodide-rich diet contributes to the activation of autoreactive cells rather than the generation of the autoantigenic epitope. Furthermore, spontaneous T4p2553-specific IgG are not detectable within the strong Tg-specific autoantibody response. To our knowledge, these data identify for the first time a Tg T cell epitope as a spontaneous target in ISAT.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Thyroglobulin from porcine thyroid gland, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
L-Lysine monohydrochloride, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium iodide, Vetec, reagent grade
Sigma-Aldrich
Sodium iodide, puriss. p.a., ≥99.0% (AT)
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%