  • Search Results
  • Adsorption of aqueous organic mixtures on a chiral stationary phase with bound antibiotic eremomycin.
Adsorption of aqueous organic mixtures on a chiral stationary phase with bound antibiotic eremomycin.
Journal of chromatography. A (2014-09-04)
Yuliya K Nikitina, Imran Ali, Leonid D Asnin
ABSTRACT
The adsorption of two typical hydro-organic mobile phases, with methanol and acetonitrile as the organic component, on an antibiotic based chiral stationary phase (CSP) Nautilus-E was studied by the minor perturbation method. In both cases, the excess adsorption of water was positive over a wide range of concentrations from 0 to ∼75 or 90 mol% for MeOH or MeCN containing mobile phases, respectively. Such hydrophilic properties of the CSP were attributed to multiple polar functional groups of the chiral ligand and to the residual silanol groups of the silica support. The adsorbed phase was found to be thinner for H₂O-MeOH (∼1.1Å) and thicker for H₂O-MeCN (∼9.4Å). The measurements of the column hold-up volume by different methods allowed us to suggest a model of the adsorbed phase consisting of the volume between bound chiral selectors inaccessible to large size molecules and of the stagnant layer of the mobile phase adsorbed on the external surface of the chiral selectors.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Water, deionized
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material