Labdane diterpenoids from Salvia reuterana.
Phytochemistry (2014-09-23)
Mahdi Moridi Farimani, Mansour Miran
ABSTRACT
Three labdane diterpenoids, 14α-hydroxy-15-chlorosclareol (1), 14α-hydroxy-15-acetoxysclareol (2), and 6β-hydroxy-14α-epoxysclareol (3), together with the known diterpenoids sclareol (4), 6β-hydroxysclareol (5), and 14α-epoxysclareol (6), as well as other common plant constituents were isolated from the n-hexane extract of aerial parts of Salvia reuterana. The structures of the new compounds were established by extensive 1D and 2D NMR spectroscopic techniques. Compound 1 is the first example of a halogenated terpenoid in the genus Salvia. Compounds 1-6 were also tested for their inhibitory activity toward HeLa and MCF-7 cell lines. Preliminary structure-activity relationship studies indicated that double bond moiety in sclareol is an essential feature for activity and modification of this moiety significantly decreased the cytotoxic activity of the resulting compounds.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sclareol, 98%
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)