  • Search Results
  • Exogenous application of methyl jasmonate lowers the effect of cadmium-induced oxidative injury in rice seedlings.
Exogenous application of methyl jasmonate lowers the effect of cadmium-induced oxidative injury in rice seedlings.
Phytochemistry (2014-10-11)
Indra Singh, Kavita Shah
ABSTRACT
Rice seedlings grown under 50 μM cadmium alone or in combination with 5 μM methyl jasmonate were investigated for Cd-induced oxidative injury at 3, 7 and 10 days of treatment. MeJA treatments alone did not have any significant change in antioxidant enzyme activities or levels of H2O2 and O2(·-) in roots/shoots, as compared to controls during 3-10 days. The Cd-stressed plants When supplemented with exogenous MeJA revealed significant and consistent changes in activities of antioxidant enzymes CAT, SOD, POD and GR paralleled with an increased GSH-pools than that in plants subjected to Cd-stress alone. Synthesis of GSH driven by increasing demand for GSH in response to Cd-induced oxidative stress in rice was evident. Increased activity of LOX under Cd-stress was noted. Results suggest enhanced Cd-tolerance, lowered Cd(2+) uptake, an improved membrane integrity and 'switching on' of the JA-biosynthesis by LOX in the Cd-stressed rice roots/shoots exposed to MeJA. Exposure to MeJA improved antioxidant response and accumulation of antioxidants which perhaps lowered the Cd-induced oxidative stress in rice. It is this switching on/off of the JA-biosynthesis and ROS mediated signal transduction pathway involving glutathione homeostasis via GR which helps MeJA to mitigate Cd-induced oxidative injury in rice.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)