  • Search Results
  • Current status of bioresorbable scaffolds in the treatment of coronary artery disease.
Current status of bioresorbable scaffolds in the treatment of coronary artery disease.
Journal of the American College of Cardiology (2014-12-17)
Jens Wiebe, Holger M Nef, Christian W Hamm
ABSTRACT
State-of-the-art drug-eluting metal stents are the gold standard for interventional treatment of coronary artery disease. Although they overcome some disadvantages and limitations of plain balloon angioplasty and bare-metal stents, some limitations apply, most notably a chronic local inflammatory reaction due to permanent implantation of a foreign body, restriction of vascular vasomotion due to a metal cage, and the risk of late and very late stent thrombosis. The development of biodegradable scaffolds is a new approach that attempts to circumvent these drawbacks. These devices provide short-term scaffolding of the vessel and then dissolve, which should theoretically circumvent the side effects of metal drug-eluting stents. Various types of these bioresorbable scaffolds are currently under clinical evaluation. This review discusses different concepts of bioresorbable scaffolds with respect to material, design, and drug elution and presents the most recent evidence.
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
Polylactic acid, Mw ~60,000
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Lactic acid solution, Vetec, reagent grade, 85%
USP
Lactic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, ReagentPlus®, powder, −50 mesh, ≥99%
Sigma-Aldrich
Magnesium, ReagentPlus®, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Magnesium, ribbon, ≥99.5% Mg basis
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, microfoil, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%